Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTGAAGCTCCTAGAGGCTAAAT[G/T]CCAGGGGGATGGAGTGAGCTACGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606075 MIM: 610454 MIM: 611848 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf2 - chromosome 10 open reading frame 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163812.1 | 851 | Missense Mutation | TGC,TTC | C,F 226 | NP_001157284.1 | |
NM_001163813.1 | 851 | Intron | NP_001157285.1 | |||
NM_001163814.1 | 851 | Intron | NP_001157286.1 | |||
NM_021830.4 | 851 | Missense Mutation | TGC,TTC | C,F 226 | NP_068602.2 | |
XM_011539975.2 | 851 | Intron | XP_011538277.1 | |||
XM_017016437.1 | 851 | UTR 5 | XP_016871926.1 |
LZTS2 - leucine zipper tumor suppressor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL43 - mitochondrial ribosomal protein L43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA4G - semaphorin 4G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |