Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCATCCACCTGGCAGAAGGCAAA[C/T]TCTGCAAAGACAGGAGAAAGGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611800 MIM: 612053 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01126 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01126 - long intergenic non-protein coding RNA 1126 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THADA - THADA, armadillo repeat containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083953.1 | 5650 | Silent Mutation | GAA,GAG | E,E 1766 | NP_001077422.1 | |
NM_001271643.1 | 5650 | Intron | NP_001258572.1 | |||
NM_001271644.1 | 5650 | Intron | NP_001258573.1 | |||
NM_022065.4 | 5650 | Silent Mutation | GAA,GAG | E,E 1766 | NP_071348.3 | |
XM_006712061.3 | 5650 | Silent Mutation | GAA,GAG | E,E 1766 | XP_006712124.1 | |
XM_006712062.1 | 5650 | Silent Mutation | GAA,GAG | E,E 1765 | XP_006712125.1 | |
XM_006712063.1 | 5650 | Silent Mutation | GAA,GAG | E,E 1726 | XP_006712126.1 | |
XM_006712064.2 | 5650 | Silent Mutation | GAA,GAG | E,E 1722 | XP_006712127.1 | |
XM_006712065.1 | 5650 | Silent Mutation | GAA,GAG | E,E 1692 | XP_006712128.1 | |
XM_006712066.1 | 5650 | Silent Mutation | GAA,GAG | E,E 1645 | XP_006712129.1 | |
XM_006712067.1 | 5650 | Silent Mutation | GAA,GAG | E,E 1644 | XP_006712130.1 | |
XM_006712068.3 | 5650 | Intron | XP_006712131.1 | |||
XM_006712069.3 | 5650 | Silent Mutation | GAA,GAG | E,E 949 | XP_006712132.1 | |
XM_011533015.2 | 5650 | Intron | XP_011531317.1 | |||
XM_017004674.1 | 5650 | Silent Mutation | GAA,GAG | E,E 1725 | XP_016860163.1 | |
XM_017004675.1 | 5650 | Silent Mutation | GAA,GAG | E,E 870 | XP_016860164.1 |
ZFP36L2 - ZFP36 ring finger protein like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |