Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAGCAGGAGTGCCTGGTCGAGA[C/T]GGGAGCCCTGGGGCCAATGGCATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610635 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTHRC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTHRC1 - collagen triple helix repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256099.1 | 305 | Silent Mutation | GAC,GAT | D,D 52 | NP_001243028.1 | |
NM_138455.3 | 305 | Silent Mutation | GAC,GAT | D,D 66 | NP_612464.1 | |
XM_011516824.2 | 305 | Silent Mutation | GAC,GAT | D,D 66 | XP_011515126.1 |