Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATTTCAGAAGGTGATAATGTGG[A/G]AAGCTGGATGCTACTGGGATGTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZCCHC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZCCHC7 - zinc finger CCHC-type containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289119.1 | 694 | Missense Mutation | GAA,GGA | E,G 174 | NP_001276048.1 | |
NM_001289120.1 | 694 | Missense Mutation | GAA,GGA | E,G 174 | NP_001276049.1 | |
NM_001289121.1 | 694 | Missense Mutation | GAA,GGA | E,G 174 | NP_001276050.1 | |
NM_032226.2 | 694 | Missense Mutation | GAA,GGA | E,G 174 | NP_115602.2 | |
XM_005251608.4 | 694 | Missense Mutation | GAA,GGA | E,G 174 | XP_005251665.1 | |
XM_005251612.2 | 694 | Missense Mutation | GAA,GGA | E,G 174 | XP_005251669.1 | |
XM_011518050.1 | 694 | Missense Mutation | GAA,GGA | E,G 174 | XP_011516352.1 | |
XM_011518051.2 | 694 | Intron | XP_011516353.1 | |||
XM_017015197.1 | 694 | Missense Mutation | GAA,GGA | E,G 174 | XP_016870686.1 | |
XM_017015198.1 | 694 | Missense Mutation | GAA,GGA | E,G 174 | XP_016870687.1 | |
XM_017015199.1 | 694 | UTR 5 | XP_016870688.1 | |||
XM_017015200.1 | 694 | UTR 5 | XP_016870689.1 | |||
XM_017015201.1 | 694 | Intron | XP_016870690.1 |