Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTGGATGCCCTGGGGCTGAGAG[G/T]AATCTTCTAGTTTACTCTTATTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF431 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF431 - zinc finger protein 431 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319124.1 | 240 | Missense Mutation | AGG,AGT | R,S 22 | NP_001306053.1 | |
NM_001319126.1 | 240 | UTR 5 | NP_001306055.1 | |||
NM_001319127.1 | 240 | UTR 5 | NP_001306056.1 | |||
XM_011527750.2 | 240 | Intron | XP_011526052.1 |