Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGGTGGTCCGGGGCCTAGGGG[C/G]ATGCCTGACCAACAGAGGCTCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611510 MIM: 607720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPT - centromere protein T | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025082.3 | 1725 | Missense Mutation | CCC,GCC | P,A 441 | NP_079358.3 | |
XM_006721277.2 | 1725 | Missense Mutation | CCC,GCC | P,A 338 | XP_006721340.1 | |
XM_011523344.1 | 1725 | Missense Mutation | CCC,GCC | P,A 338 | XP_011521646.1 | |
XM_011523345.1 | 1725 | Missense Mutation | CCC,GCC | P,A 194 | XP_011521647.1 | |
XM_017023712.1 | 1725 | Missense Mutation | CCC,GCC | P,A 338 | XP_016879201.1 | |
XM_017023713.1 | 1725 | Missense Mutation | CCC,GCC | P,A 199 | XP_016879202.1 | |
XM_017023714.1 | 1725 | Missense Mutation | CCC,GCC | P,A 199 | XP_016879203.1 | |
XM_017023715.1 | 1725 | Missense Mutation | CCC,GCC | P,A 199 | XP_016879204.1 | |
XM_017023716.1 | 1725 | Missense Mutation | CCC,GCC | P,A 194 | XP_016879205.1 | |
XM_017023717.1 | 1725 | Missense Mutation | CCC,GCC | P,A 194 | XP_016879206.1 |