Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCTGGGTGCCTGTGCTGTGGTG[G/T]GGCCATTCCATGGCCCTGAGTGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608797 MIM: 601304 | ||||||||||||||||||||
Literature Links: |
C22orf46 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C22orf46 - chromosome 22 open reading frame 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142964.1 | 122 | Missense Mutation | GGG,TGG | G,W 13 | NP_001136436.1 | |
XM_011530373.2 | 122 | Missense Mutation | GGG,TGG | G,W 13 | XP_011528675.1 | |
XM_017028939.1 | 122 | Intron | XP_016884428.1 |
MEI1 - meiotic double-stranded break formation protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNU13 - SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003796.1 | 122 | Intron | NP_001003796.1 | |||
NM_005008.3 | 122 | Intron | NP_004999.1 | |||
XM_017028811.1 | 122 | Intron | XP_016884300.1 |