Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAAACCTGTCTTGAAAAGAACA[A/C]TGGCTATCATCATCCTCTGTCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604775 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSC-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSC-AS1 - MSC antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRPA1 - transient receptor potential cation channel subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007332.2 | 3427 | Missense Mutation | CAG,CAT | Q,H 1084 | NP_015628.2 | |
XM_011517624.2 | 3427 | Missense Mutation | CAG,CAT | Q,H 1109 | XP_011515926.1 | |
XM_011517625.2 | 3427 | Missense Mutation | CAG,CAT | Q,H 1084 | XP_011515927.1 | |
XM_017013946.1 | 3427 | Missense Mutation | CAG,CAT | Q,H 1084 | XP_016869435.1 |