Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCGGGAACACTCAATCTCCTGG[C/T]GCAGGAGAGCAAGATGGTGCTTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610648 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUX2 - cut like homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015267.3 | Intron | NP_056082.2 | ||||
XM_011538061.2 | Intron | XP_011536363.1 | ||||
XM_011538063.2 | Intron | XP_011536365.1 | ||||
XM_011538069.2 | Intron | XP_011536371.1 | ||||
XM_011538070.2 | Intron | XP_011536372.1 | ||||
XM_017019080.1 | Intron | XP_016874569.1 | ||||
XM_017019081.1 | Intron | XP_016874570.1 |