Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATTTAGGCCGAATGAACTACCA[C/T]GAGAACAGCCACATACCTCACTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608183 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHSY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHSY1 - chondroitin sulfate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014918.4 | 3020 | UTR 3 | NP_055733.2 | |||
XM_006720435.3 | 3020 | UTR 3 | XP_006720498.1 | |||
XM_011521364.2 | 3020 | UTR 3 | XP_011519666.1 | |||
XM_017022011.1 | 3020 | UTR 3 | XP_016877500.1 |