Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCCGCGCCCGCGAGATCTTCAC[C/T]TCGCTGGAGTACGGACCGGTGCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613358 MIM: 611480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH16A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH16A1 - aldehyde dehydrogenase 16 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145396.1 | 161 | Silent Mutation | ACC,ACT | T,T 15 | NP_001138868.1 | |
NM_153329.3 | 161 | Silent Mutation | ACC,ACT | T,T 15 | NP_699160.2 | |
XM_011526441.1 | 161 | Intron | XP_011524743.1 | |||
XM_011526442.1 | 161 | Intron | XP_011524744.1 |
PIH1D1 - PIH1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017916.2 | 161 | Intron | NP_060386.1 |