Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAAAGCAGTCGTTATGCCACTGC[C/T]GTTCCTCAAAGGAGATGTATTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602633 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf49 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf49 - chromosome 2 open reading frame 49 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FHL2 - four and a half LIM domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039492.2 | 949 | Missense Mutation | NP_001034581.1 | |||
NM_001318894.1 | 949 | Missense Mutation | NP_001305823.1 | |||
NM_001318895.1 | 949 | Missense Mutation | NP_001305824.1 | |||
NM_001318896.1 | 949 | Missense Mutation | NP_001305825.1 | |||
NM_001318897.1 | 949 | Missense Mutation | NP_001305826.1 | |||
NM_001318898.1 | 949 | Missense Mutation | NP_001305827.1 | |||
NM_001318899.1 | 949 | Missense Mutation | NP_001305828.1 | |||
NM_001450.3 | 949 | Missense Mutation | NP_001441.4 | |||
NM_201555.1 | 949 | Missense Mutation | NP_963849.1 | |||
NM_201557.3 | 949 | Missense Mutation | NP_963851.2 | |||
XM_011510798.2 | 949 | Missense Mutation | XP_011509100.1 |