Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAAACAAGTATTCCTGATTTGG[A/T]GTAAGGTCTAGATACTGAACGATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM209 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM209 - transmembrane protein 209 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301163.1 | 1625 | Intron | NP_001288092.1 | |||
NM_032842.3 | 1625 | Silent Mutation | ACA,ACT | T,T 404 | NP_116231.2 | |
XM_005250655.1 | 1625 | Silent Mutation | ACA,ACT | T,T 403 | XP_005250712.1 | |
XM_011516646.2 | 1625 | Silent Mutation | ACA,ACT | T,T 403 | XP_011514948.1 |