Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGGAGAAGACCCTCCTCTCCT[A/C]CTCCCATCCTCTAGTGCCTGGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300762 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGEB16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGEB16 - MAGE family member B16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099921.1 | 290 | Missense Mutation | TAC,TCC | Y,S 41 | NP_001093391.1 | |
XM_011545453.2 | 290 | Missense Mutation | TAC,TCC | Y,S 41 | XP_011543755.1 | |
XM_011545454.2 | 290 | Missense Mutation | TAC,TCC | Y,S 41 | XP_011543756.1 |