Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGTTTCTGAAATACCACCAGGC[A/G]TGGAGCTGGGCCACCAACGGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613699 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLT6D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLT6D1 - glycosyltransferase 6 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182974.2 | 1163 | Silent Mutation | CAC,CAT | H,H 195 | NP_892019.2 | |
XM_011518633.1 | 1163 | Silent Mutation | CAC,CAT | H,H 231 | XP_011516935.1 | |
XM_011518634.1 | 1163 | Silent Mutation | CAC,CAT | H,H 231 | XP_011516936.1 | |
XM_011518635.1 | 1163 | Silent Mutation | CAC,CAT | H,H 231 | XP_011516937.1 | |
XM_011518636.1 | 1163 | Silent Mutation | CAC,CAT | H,H 231 | XP_011516938.1 | |
XM_011518637.1 | 1163 | Silent Mutation | CAC,CAT | H,H 195 | XP_011516939.1 | |
XM_011518638.2 | 1163 | Silent Mutation | CAC,CAT | H,H 163 | XP_011516940.1 |
LOC102723971 - lipocalin 1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |