Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGTGCAGACGCTGGCCCGGAT[A/G]TCGCGGAGGACTCGCGGGCCGGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf223 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf223 - chromosome 6 open reading frame 223 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171992.2 | 1441 | UTR 3 | NP_001165463.1 | |||
NM_001324369.1 | 1441 | Missense Mutation | ATA,ATG | I,M 1 | NP_001311298.1 | |
NM_153246.5 | 1441 | Missense Mutation | ATA,ATG | I,M 113 | NP_694978.2 | |
XM_017010425.1 | 1441 | Missense Mutation | ATA,ATG | I,M 112 | XP_016865914.1 |
LOC101929705 - uncharacterized LOC101929705 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |