Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCCACATGAGCCAGAAACACAC[A/G]GAGATGGTGAGAGGTGTGGGATGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 112264 MIM: 608302 MIM: 178620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMP1 - bone morphogenetic protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LGI3 - leucine rich repeat LGI family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFTPC - surfactant protein C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172357.1 | 353 | Silent Mutation | ACA,ACG | T,T 65 | NP_001165828.1 | |
NM_001172410.1 | 353 | Silent Mutation | ACA,ACG | T,T 65 | NP_001165881.1 | |
NM_001317778.1 | 353 | Silent Mutation | ACA,ACG | T,T 65 | NP_001304707.1 | |
NM_001317779.1 | 353 | Intron | NP_001304708.1 | |||
NM_001317780.1 | 353 | Silent Mutation | ACA,ACG | T,T 65 | NP_001304709.1 | |
NM_003018.3 | 353 | Silent Mutation | ACA,ACG | T,T 65 | NP_003009.2 | |
XM_011544613.2 | 353 | Intron | XP_011542915.1 |