Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAAGGTGAGAGAGGGCATAAGTA[C/T]GGTGACTGTAGCCCCCGAGCACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL33 - kelch like family member 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109997.2 | 2649 | Missense Mutation | CAT,CGT | H,R 476 | NP_001103467.2 | |
XM_005267347.4 | 2649 | Missense Mutation | CAT,CGT | H,R 740 | XP_005267404.1 | |
XM_011536450.2 | 2649 | Missense Mutation | CAT,CGT | H,R 740 | XP_011534752.1 |