Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGGCAAGATGCGAACACCACAG[A/C]CATCTAGTAAACTTCGGGAATCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C7orf50 - chromosome 7 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134395.1 | Intron | NP_001127867.1 | ||||
NM_001134396.1 | Intron | NP_001127868.1 | ||||
NM_001318252.1 | Intron | NP_001305181.1 | ||||
NM_032350.5 | Intron | NP_115726.1 | ||||
XM_006715793.2 | Intron | XP_006715856.1 | ||||
XM_011515580.2 | Intron | XP_011513882.1 | ||||
XM_011515581.2 | Intron | XP_011513883.1 | ||||
XM_011515582.2 | Intron | XP_011513884.1 | ||||
XM_011515583.2 | Intron | XP_011513885.1 | ||||
XM_011515584.2 | Intron | XP_011513886.1 | ||||
XM_017012719.1 | Intron | XP_016868208.1 | ||||
XM_017012720.1 | Intron | XP_016868209.1 | ||||
XM_017012721.1 | Intron | XP_016868210.1 | ||||
XM_017012722.1 | Intron | XP_016868211.1 |
GPR146 - G protein-coupled receptor 146 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303473.1 | Intron | NP_001290402.1 | ||||
NM_001303474.1 | Intron | NP_001290403.1 | ||||
NM_138445.3 | Intron | NP_612454.1 | ||||
XM_005249613.2 | Intron | XP_005249670.1 | ||||
XM_006715642.2 | Intron | XP_006715705.1 | ||||
XM_006715643.2 | Intron | XP_006715706.1 | ||||
XM_011515116.2 | Intron | XP_011513418.1 |
LOC107986755 - uncharacterized LOC107986755 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |