Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATTTCCTGATAACAGGTCCCAA[A/G]GTAGGATGATCTCCAGCTTCTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WNT8A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WNT8A - Wnt family member 8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300938.1 | 370 | Silent Mutation | AAA,AAG | K,K 52 | NP_001287867.1 | |
NM_001300939.1 | 370 | Silent Mutation | AAA,AAG | K,K 52 | NP_001287868.1 | |
NM_058244.3 | 370 | Silent Mutation | AAA,AAG | K,K 34 | NP_490645.1 | |
XM_011543625.2 | 370 | Silent Mutation | AAA,AAG | K,K 74 | XP_011541927.2 | |
XM_017009824.1 | 370 | Silent Mutation | AAA,AAG | K,K 74 | XP_016865313.1 | |
XM_017009825.1 | 370 | Silent Mutation | AAA,AAG | K,K 74 | XP_016865314.1 | |
XM_017009826.1 | 370 | UTR 5 | XP_016865315.1 |