Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTCGTCCGAACCTTCAGGGAGC[A/G]CGAGGTGCCCTGGGGAAAGGGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602302 MIM: 608302 MIM: 609349 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HR - hair growth associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LGI3 - leucine rich repeat LGI family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REEP4 - receptor accessory protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316964.1 | 1032 | Missense Mutation | NP_001303893.1 | |||
NM_001316965.1 | 1032 | UTR 3 | NP_001303894.1 | |||
NM_025232.3 | 1032 | Missense Mutation | NP_079508.2 |