Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGAAGGAAGGTAAAACTGTTAA[G/T]GGTGAGCTTAAGTAATACACAGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614762 | ||||||||||||||||||||
Literature Links: |
GLYATL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLYATL2 - glycine-N-acyltransferase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145016.3 | 1146 | UTR 3 | NP_659453.3 | |||
XM_011544809.2 | 1146 | Intron | XP_011543111.1 | |||
XM_017017337.1 | 1146 | UTR 3 | XP_016872826.1 | |||
XM_017017338.1 | 1146 | UTR 3 | XP_016872827.1 |