Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCTCACACTCTCCAACCTCAT[C/T]TCCCTGGTGAGAGGCCCTCCGGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100720 MIM: 613858 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRND PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRND - cholinergic receptor nicotinic delta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000751.2 | 220 | Silent Mutation | ATC,ATT | I,I 64 | NP_000742.1 | |
NM_001256657.1 | 220 | Silent Mutation | ATC,ATT | I,I 64 | NP_001243586.1 | |
NM_001311195.1 | 220 | UTR 5 | NP_001298124.1 | |||
NM_001311196.1 | 220 | UTR 5 | NP_001298125.1 | |||
XM_011510524.2 | 220 | UTR 5 | XP_011508826.1 |
PRSS56 - protease, serine 56 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195129.1 | 220 | Intron | NP_001182058.1 |