Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCACCGGCAATGGAGCGGTTGAC[A/G]TTGCCTCTCGGCGGCGCGGCGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM221A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM221A - family with sequence similarity 221 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127364.2 | 115 | Silent Mutation | ACA,ACG | T,T 5 | NP_001120836.1 | |
NM_001127365.2 | 115 | Silent Mutation | ACA,ACG | T,T 5 | NP_001120837.1 | |
NM_001300932.1 | 115 | Silent Mutation | ACA,ACG | T,T 5 | NP_001287861.1 | |
NM_199136.4 | 115 | Silent Mutation | ACA,ACG | T,T 5 | NP_954587.2 | |
XM_011515369.2 | 115 | Silent Mutation | ACA,ACG | T,T 5 | XP_011513671.1 | |
XM_011515370.2 | 115 | Silent Mutation | ACA,ACG | T,T 5 | XP_011513672.1 | |
XM_011515371.2 | 115 | Silent Mutation | ACA,ACG | T,T 5 | XP_011513673.1 | |
XM_011515372.2 | 115 | Intron | XP_011513674.1 | |||
XM_017012134.1 | 115 | Silent Mutation | ACA,ACG | T,T 5 | XP_016867623.1 | |
XM_017012135.1 | 115 | Intron | XP_016867624.1 |