Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTACGAGGTGCACCCCAACCGA[C/T]GCCCCAAGATCCTGGCCCAGACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609831 MIM: 176763 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC163P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC163P - coiled-coil domain containing 163, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMACHC - methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015506.2 | 392 | Missense Mutation | CGC,TGC | R,C 112 | NP_056321.2 | |
XM_005270724.4 | 392 | Missense Mutation | CGC,TGC | R,C 47 | XP_005270781.1 | |
XM_011541204.2 | 392 | Missense Mutation | CGC,TGC | R,C 55 | XP_011539506.1 |
PRDX1 - peroxiredoxin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |