Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAAAACAAAACAAAAAAAAGAA[A/G]CAACAACAAAACCCAAACTATTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164035 MIM: 604012 MIM: 611133 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCL - nucleolin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005381.2 | 2581 | UTR 3 | NP_005372.2 |
SNORA75 - small nucleolar RNA, H/ACA box 75 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD20 - small nucleolar RNA, C/D box 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD82 - small nucleolar RNA, C/D box 82 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |