Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGTATCTGGAAGTTATCAATACC[A/G]TGGCACAGGTCACTTTTGACATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615843 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEGS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEGS1 - delta 4-desaturase, sphingolipid 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321541.1 | 700 | Missense Mutation | ATG,GTG | M,V 191 | NP_001308470.1 | |
NM_001321542.1 | 700 | Missense Mutation | ATG,GTG | M,V 155 | NP_001308471.1 | |
NM_003676.3 | 700 | Missense Mutation | ATG,GTG | M,V 191 | NP_003667.1 | |
XM_017002648.1 | 700 | Missense Mutation | ATG,GTG | M,V 155 | XP_016858137.1 |