Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGTTAGTACAGATCCCAATATT[C/T]TTTTTCACTGTTGTAGTAATGTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606821 MIM: 601910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COG5 - component of oligomeric golgi complex 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161520.1 | 2006 | Intron | NP_001154992.1 | |||
NM_006348.3 | 2006 | Intron | NP_006339.3 | |||
NM_181733.2 | 2006 | Intron | NP_859422.2 |
GPR22 - G protein-coupled receptor 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005295.2 | 2006 | Silent Mutation | TTC,TTT | F,F 220 | NP_005286.2 | |
XM_011516056.2 | 2006 | Silent Mutation | TTC,TTT | F,F 220 | XP_011514358.1 |