Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAGAGCTTGCAAGTTTGCTTCAC[A/G]GTTCAAAAATCCCATTGCACTGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605046 | ||||||||||||||||||||
Literature Links: |
UBQLN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBQLN1 - ubiquilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013438.4 | 2174 | Missense Mutation | CGT,TGT | R,C 562 | NP_038466.2 | |
NM_053067.2 | 2174 | Missense Mutation | CGT,TGT | R,C 534 | NP_444295.1 | |
XM_005251948.2 | 2174 | Intron | XP_005252005.1 |