Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGAAGAGAGAGCTGACAAAAGG[C/T]TTGTGCTCAGGTTAGTGAAGAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF569 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF569 - zinc finger protein 569 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF570 - zinc finger protein 570 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300993.2 | 284 | Silent Mutation | GGC,GGT | G,G 138 | NP_001287922.1 | |
NM_001321991.1 | 284 | Silent Mutation | GGC,GGT | G,G 82 | NP_001308920.1 | |
NM_001321992.1 | 284 | Silent Mutation | GGC,GGT | G,G 40 | NP_001308921.1 | |
NM_001321993.1 | 284 | UTR 5 | NP_001308922.1 | |||
NM_001321994.1 | 284 | UTR 5 | NP_001308923.1 | |||
NM_144694.3 | 284 | Silent Mutation | GGC,GGT | G,G 82 | NP_653295.1 | |
XM_011526544.2 | 284 | Silent Mutation | GGC,GGT | G,G 82 | XP_011524846.1 | |
XM_011526545.2 | 284 | Silent Mutation | GGC,GGT | G,G 40 | XP_011524847.1 |