Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGACGGGCAGCCCTGAGCTCCAG[C/T]TGCAGTTCTCCAATGATATCTACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610534 | ||||||||||||||||||||
Literature Links: |
DCPS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCPS - decapping enzyme, scavenger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014026.4 | 1096 | Silent Mutation | CTG,TTG | L,L 106 | NP_054745.1 | |
XM_011542778.2 | 1096 | Silent Mutation | CTG,TTG | L,L 113 | XP_011541080.1 | |
XM_011542780.2 | 1096 | Intron | XP_011541082.1 | |||
XM_017017612.1 | 1096 | UTR 5 | XP_016873101.1 |
HP11014 - uncharacterized HP11014 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |