Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGTGCGGAACCGCTCCTGGCCG[A/G]CCGTGTCCCAGATCTGCAGCTAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ISY1-RAB43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ISY1-RAB43 - ISY1-RAB43 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204890.1 | 525 | Missense Mutation | CCG,TCG | P,S 291 | NP_001191819.1 |
RAB43 - RAB43, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204883.1 | 525 | Intron | NP_001191812.1 | |||
NM_001204884.1 | 525 | Intron | NP_001191813.1 | |||
NM_001204885.1 | 525 | Intron | NP_001191814.1 | |||
NM_001204886.1 | 525 | Intron | NP_001191815.1 | |||
NM_001204887.1 | 525 | Missense Mutation | GCC,GTC | A,V 75 | NP_001191816.1 | |
NM_001204888.1 | 525 | Silent Mutation | GGC,GGT | G,G 70 | NP_001191817.1 | |
NM_198490.2 | 525 | Missense Mutation | GCC,GTC | A,V 75 | NP_940892.1 |