Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGATGGACACGTCCCTGAGGGC[A/G]ATGGGGGGCACCTCCAGCCAGGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614781 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TECPR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TECPR1 - tectonin beta-propeller repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015395.2 | 3096 | Silent Mutation | ATC,ATT | I,I 932 | NP_056210.1 | |
XM_005250253.3 | 3096 | Silent Mutation | ATC,ATT | I,I 932 | XP_005250310.1 | |
XM_017011937.1 | 3096 | Silent Mutation | ATC,ATT | I,I 898 | XP_016867426.1 | |
XM_017011938.1 | 3096 | Intron | XP_016867427.1 |