Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTACCCGTATAATGAAGGAAAC[A/G]TGAAAGATGTTTTCCACTGTACGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612987 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATE1-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATE1-AS1 - ATE1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NSMCE4A - NSE4 homolog A, SMC5-SMC6 complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167865.1 | 984 | Silent Mutation | CAC,CAT | H,H 307 | NP_001161337.1 | |
NM_017615.2 | 984 | Silent Mutation | CAC,CAT | H,H 307 | NP_060085.2 | |
XM_005269929.4 | 984 | Silent Mutation | CAC,CAT | H,H 148 | XP_005269986.1 | |
XM_005269930.3 | 984 | Silent Mutation | CAC,CAT | H,H 148 | XP_005269987.1 | |
XM_011539910.2 | 984 | Silent Mutation | CAC,CAT | H,H 148 | XP_011538212.1 | |
XM_017016359.1 | 984 | Silent Mutation | CAC,CAT | H,H 148 | XP_016871848.1 |