Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGACTCACTCGAGGCTGACAGC[A/G]GGGGTAGCTAAACAGGTGAACTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EML2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EML2 - echinoderm microtubule associated protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193268.1 | 2567 | Missense Mutation | NP_001180197.1 | |||
NM_001193269.1 | 2567 | Missense Mutation | NP_001180198.1 | |||
NM_012155.2 | 2567 | Missense Mutation | NP_036287.1 | |||
XM_006723125.2 | 2567 | Missense Mutation | XP_006723188.1 | |||
XM_006723126.2 | 2567 | Missense Mutation | XP_006723189.1 | |||
XM_011526694.2 | 2567 | Missense Mutation | XP_011524996.1 | |||
XM_011526695.2 | 2567 | Missense Mutation | XP_011524997.1 | |||
XM_011526698.2 | 2567 | Missense Mutation | XP_011525000.1 | |||
XM_011526700.2 | 2567 | Missense Mutation | XP_011525002.1 | |||
XM_011526701.2 | 2567 | Missense Mutation | XP_011525003.1 | |||
XM_017026550.1 | 2567 | Missense Mutation | XP_016882039.1 | |||
XM_017026551.1 | 2567 | Intron | XP_016882040.1 |