Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATCTACACCATTGAAAGCTGCA[A/G]AACCGTTGAGGTGCTGAGCAGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603728 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUMB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUMB - NUMB, endocytic adaptor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005743.1 | 2044 | Missense Mutation | TCT,TTT | S,F 586 | NP_001005743.1 | |
NM_001005744.1 | 2044 | Missense Mutation | TCT,TTT | S,F 538 | NP_001005744.1 | |
NM_001005745.1 | 2044 | Missense Mutation | TCT,TTT | S,F 527 | NP_001005745.1 | |
NM_001320114.1 | 2044 | Missense Mutation | TCT,TTT | S,F 538 | NP_001307043.1 | |
NM_003744.5 | 2044 | Missense Mutation | TCT,TTT | S,F 575 | NP_003735.3 |
PAPLN - papilin, proteoglycan like sulfated glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |