Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTCTAACTGAAGCAGCCTGGGA[A/G]TATCTGGGTACCCACCCAGCTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM186A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM186A - family with sequence similarity 186 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145475.1 | 7105 | Missense Mutation | ACT,ATT | T,I 2323 | NP_001138947.1 | |
XM_006719231.3 | 7105 | Missense Mutation | ACT,ATT | T,I 2323 | XP_006719294.1 | |
XM_011537890.2 | 7105 | Missense Mutation | ACT,ATT | T,I 2319 | XP_011536192.1 | |
XM_011537892.1 | 7105 | Missense Mutation | ACT,ATT | T,I 2180 | XP_011536194.1 |