Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCTTGTTCCTCTTCTCCTTCG[C/T]CTTTGGTGTTCTCTTCCCTGCAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSPO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RSPO2 - R-spondin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282863.1 | 1278 | Missense Mutation | ACG,GCG | T,A 148 | NP_001269792.1 | |
NM_001317942.1 | 1278 | Missense Mutation | ACG,GCG | T,A 145 | NP_001304871.1 | |
NM_178565.4 | 1278 | Missense Mutation | ACG,GCG | T,A 212 | NP_848660.3 | |
XM_011517018.1 | 1278 | Missense Mutation | ACG,GCG | T,A 219 | XP_011515320.1 | |
XM_011517019.1 | 1278 | Missense Mutation | ACG,GCG | T,A 218 | XP_011515321.1 | |
XM_017013395.1 | 1278 | Missense Mutation | ACG,GCG | T,A 211 | XP_016868884.1 |