Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTTCTTCACTTGCCAAAATATC[C/T]ACTGGGTCTTCAGCAATTTCTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615488 MIM: 615487 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KANSL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KANSL2 - KAT8 regulatory NSL complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017822.3 | 1413 | Silent Mutation | NP_060292.3 |
MIR1291 - microRNA 1291 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA2A - small nucleolar RNA, H/ACA box 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA2C - small nucleolar RNA, H/ACA box 2C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |