Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCAGGCCCCGCGGGAAGCGGGGG[A/G]AGTAGTCGGAGATCTGCTTGATGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604568 | ||||||||||||||||||||
Literature Links: |
DOC2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DOC2B - double C2 domain beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003585.4 | 261 | Missense Mutation | TCC,TTC | S,F 37 | NP_003576.2 | |
XM_011535363.2 | 261 | Missense Mutation | TCC,TTC | S,F 37 | XP_011533665.1 | |
XM_017025227.1 | 261 | Missense Mutation | TCC,TTC | S,F 37 | XP_016880716.1 |
LOC100506371 - uncharacterized LOC100506371 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |