Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGATTCTGAAAGGCAGTGTGAC[A/G]TTTCTTTCTCCACCTAGGGATGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGCT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGCT - gamma-glutamylcyclotransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199815.1 | Intron | NP_001186744.1 | ||||
NM_001199816.1 | Intron | NP_001186745.1 | ||||
NM_001199817.1 | Intron | NP_001186746.1 | ||||
NM_024051.3 | Intron | NP_076956.1 |