Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACGGATGCTTCAAAAAGTGACT[A/G]TGAACTTCAAGCTTTAAGAAATGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC160 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC160 - coiled-coil domain containing 160 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101357.1 | 791 | Missense Mutation | TAT,TGT | Y,C 201 | NP_001094827.1 | |
XM_005262409.4 | 791 | Missense Mutation | TAT,TGT | Y,C 201 | XP_005262466.1 | |
XM_005262410.4 | 791 | Missense Mutation | TAT,TGT | Y,C 201 | XP_005262467.1 |