Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGGGCAATGTCCAGGCCGAGC[A/T]GCGTCTCCCCGCGGCAGCCGGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
YTHDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YTHDC2 - YTH domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022828.3 | 229 | Missense Mutation | AGC,TGC | S,C 6 | NP_073739.3 | |
XM_011543581.1 | 229 | Missense Mutation | AGC,TGC | S,C 6 | XP_011541883.1 | |
XM_011543582.2 | 229 | Missense Mutation | AGC,TGC | S,C 6 | XP_011541884.1 | |
XM_011543586.2 | 229 | Missense Mutation | AGC,TGC | S,C 6 | XP_011541888.1 | |
XM_017009730.1 | 229 | Missense Mutation | AGC,TGC | S,C 6 | XP_016865219.1 | |
XM_017009731.1 | 229 | UTR 5 | XP_016865220.1 | |||
XM_017009732.1 | 229 | UTR 5 | XP_016865221.1 | |||
XM_017009733.1 | 229 | UTR 5 | XP_016865222.1 | |||
XM_017009734.1 | 229 | UTR 5 | XP_016865223.1 | |||
XM_017009735.1 | 229 | UTR 5 | XP_016865224.1 |