Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGGCCCGCCAGCTCTGGGGCTG[C/T]CTCTCCTTGTCCTCTTCAGAGCTCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609739 | |||||||||||||||||||||||
Literature Links: |
ILDR1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ILDR1 - immunoglobulin like domain containing receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199799.1 | 1406 | Silent Mutation | AGA,AGG | R,R 487 | NP_001186728.1 | |
NM_001199800.1 | 1406 | Silent Mutation | AGA,AGG | R,R 398 | NP_001186729.1 | |
NM_175924.3 | 1406 | Silent Mutation | AGA,AGG | R,R 443 | NP_787120.1 | |
XM_005247389.4 | 1406 | Silent Mutation | AGA,AGG | R,R 455 | XP_005247446.1 | |
XM_011512738.2 | 1406 | Silent Mutation | AGA,AGG | R,R 487 | XP_011511040.1 | |
XM_011512739.2 | 1406 | Silent Mutation | AGA,AGG | R,R 308 | XP_011511041.1 |