Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGCTATGAAGTAAATGAGCATG[C/T]CAAATGTAATGAATTTGGCTTCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613572 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPRC6A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPRC6A - G protein-coupled receptor class C group 6 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286354.1 | 2404 | Missense Mutation | GAC,GGC | D,G 612 | NP_001273283.1 | |
NM_001286355.1 | 2404 | Missense Mutation | GAC,GGC | D,G 716 | NP_001273284.1 | |
NM_148963.3 | 2404 | Missense Mutation | GAC,GGC | D,G 787 | NP_683766.2 | |
XM_017010475.1 | 2404 | Missense Mutation | GAC,GGC | D,G 740 | XP_016865964.1 | |
XM_017010476.1 | 2404 | Intron | XP_016865965.1 |