Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTATGTCTTTCTTTCCAGCTCCA[C/T]GATGCGTTATCTGGGTCTGGAAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153619 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGALS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGALS3 - lectin, galactoside binding soluble 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177388.1 | 112 | Silent Mutation | CAC,CAT | H,H 8 | NP_001170859.1 | |
NM_002306.3 | 112 | Silent Mutation | CAC,CAT | H,H 8 | NP_002297.2 | |
XM_011536759.2 | 112 | Silent Mutation | CAC,CAT | H,H 22 | XP_011535061.2 |