Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGAAAGCACAGCAGATATTGAT[G/A]TAGAAGAAAGCTTTTTAATGCAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616049 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PABPC1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PABPC1L - poly(A) binding protein cytoplasmic 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001124756.2 | 1890 | Intron | NP_001118228.1 |
TOMM34 - translocase of outer mitochondrial membrane 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006809.4 | 1890 | UTR 3 | NP_006800.2 | |||
XM_005260254.2 | 1890 | UTR 3 | XP_005260311.1 | |||
XM_011528501.1 | 1890 | UTR 3 | XP_011526803.1 | |||
XM_017027600.1 | 1890 | UTR 3 | XP_016883089.1 |