Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACAGGAATGCAGGAGTGCAGAA[A/G]GTATTTTAAAATGCTTTCGAGGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612442 | ||||||||||||||||||||
Literature Links: |
SEC22A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEC22A - SEC22 homolog A, vesicle trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012430.4 | 239 | Missense Mutation | AAG,AGG | K,R 35 | NP_036562.2 | |
XM_011512673.2 | 239 | Missense Mutation | AAG,AGG | K,R 35 | XP_011510975.1 | |
XM_011512675.2 | 239 | Missense Mutation | AAG,AGG | K,R 35 | XP_011510977.1 | |
XM_011512676.2 | 239 | Missense Mutation | AAG,AGG | K,R 35 | XP_011510978.1 |