Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTAAATTGTTGCTGGTATGAAA[A/G]TGGTCAGAGAAGCCCTCTTCCCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRMD5 - FERM domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286490.1 | 2864 | UTR 3 | NP_001273419.1 | |||
NM_001286491.1 | 2864 | UTR 3 | NP_001273420.1 | |||
NM_001322949.1 | 2864 | UTR 3 | NP_001309878.1 | |||
NM_001322950.1 | 2864 | UTR 3 | NP_001309879.1 | |||
NM_001322951.1 | 2864 | UTR 3 | NP_001309880.1 | |||
NM_032892.4 | 2864 | UTR 3 | NP_116281.2 | |||
XM_005254730.3 | 2864 | UTR 3 | XP_005254787.1 |
PIN4P1 - peptidylprolyl cis/trans isomerase, NIMA-interacting 4 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR76 - WD repeat domain 76 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |