Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGAAGTAACTGATGTGGACATG[C/G]TTTGTCAAATGTAAGGAGATACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615606 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTNL8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTNL8 - butyrophilin like 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040462.2 | 324 | Intron | NP_001035552.1 | |||
NM_001159707.1 | 324 | Intron | NP_001153179.1 | |||
NM_001159708.1 | 324 | Intron | NP_001153180.1 | |||
NM_001159709.1 | 324 | Missense Mutation | TGC,TGG | C,W 4 | NP_001153181.1 | |
NM_001159710.1 | 324 | Intron | NP_001153182.1 | |||
NM_024850.2 | 324 | Intron | NP_079126.1 | |||
XM_011534649.1 | 324 | Intron | XP_011532951.1 | |||
XM_011534650.2 | 324 | Intron | XP_011532952.1 | |||
XM_011534651.2 | 324 | Intron | XP_011532953.1 | |||
XM_011534652.2 | 324 | Intron | XP_011532954.1 | |||
XM_011534653.2 | 324 | Intron | XP_011532955.1 |